Reagents Ammonium Hydroxide, 26 Be, 2.5L, Purified

Representative image only.

Reagents Ammonium Hydroxide, 26 Be, 2.5L, Purified

Reagents | Mfr: C5103000-2.5D1| Item: UX-78919-79

Manufactured using the highest quality raw materials in ISO 9001 facility

$105.75USD / Each

$105.75USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

3 Year Warranty
Specifications
  • Purity GradePurified
  • ClassSalts
  • FormLiquid
  • Volume (mL)2500
  • Vapor Pressure (torr)580
  • PackagingClear Glass Bottle
  • DOT InformationAMMONIA SOLUTION, 8
  • UN NumberUN2672
  • CAS Number1336-21-6
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • DescriptionAmmonium Hydroxide, 26 Be, 2.5L, Purified
  • Warranty3 year
Documents & Certificates
Reviews
Q&A