Reagents Ammonium Persulfate, 100g, Purified

Representative image only.

Reagents Ammonium Persulfate, 100g, Purified

Reagents | Mfr: C2136500-100C2| Item: UX-78910-32

Manufactured using the highest quality raw materials in ISO 9001 facility

$59.00USD / Each

$59.00USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear Formula(NH4)2S2O8
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight228.2
  • PackagingAmber Jar
  • DOT InformationAMMONIUM PERSULFATE, 5.1
  • CAS Number7727-54-0
  • UN NumberUN1444
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionAmmonium Persulfate, 100g, Purified
  • Warranty7 year
Documents & Certificates
Reviews
Q&A