Reagents Ammonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 4 L

Representative image only.

Reagents Ammonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 4 L

Reagents | Mfr: CA010300-4C| Item: UX-78922-61

Manufactured using the highest quality raw materials in ISO 9001 facility

$183.50USD / Each

$183.50USD / Each
Product Description

Key Features

  • N/10
  • Assay 0.0995 to 0.1005 Normal
  • Aqueous
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 4.5 to 6
  • 4 L amber glass bottle
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Normality0.1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Shelf Life (days)760
  • Shelf Life (months)24
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionAmmonium Thiocyanate, 0.10N, Standardized, Amber Glass Bottle, 4 L
  • Warranty2 year
Documents & Certificates
Reviews
Q&A