Reagents Barium Chloride, 20% (w/v), Volumetric, Cubitainer® Package, 20 L

Representative image only.

Reagents Barium Chloride, 20% (w/v), Volumetric, Cubitainer® Package, 20 L

Reagents | Mfr: CB100500-20F| Item: UX-78929-43

Manufactured using the highest quality raw materials in ISO 9001 facility

$450.00USD / Each

$450.00USD / Each
Product Description

Key Features

  • Aqueous
  • Colorless liquid
  • Density: 1.16
  • Boiling point: approximately 101 °C (213.8 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 5.2 to 8.2
  • 20 L Cubitainer package
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

3 Year Warranty
Specifications
  • Concentration20% w/v
  • ClassVolumetric
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)20000
  • PackagingCubitainer
  • DOT InformationTOXIC LIQUID, INORGANIC, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN3287
  • Shelf Life (days)1095
  • Shelf Life (months)36
  • DescriptionBarium Chloride, 20% (w/v), Volumetric, Cubitainer® Package, 20 L
  • Warranty3 year
Documents & Certificates
Reviews
Q&A