Reagents Barium Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 100 g

Representative image only.

Reagents Barium Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 100 g

Reagents | Mfr: C1147000-100A6| Item: UX-62281-54

Manufactured using the highest quality raw materials in ISO 9001 facility

$55.80USD / Each

$55.80USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 3.86
  • Solubility information: 370 g/L at 20 °C (68 °F)
  • pH range: 5.2 to 8.2
  • 100 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaBaCl2·2H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsBarium chloride 2-hydrate
  • FormSolid
  • Weight (g)100
  • Formula Weight244.26
  • PackagingWide-mouth container
  • DOT InformationBARIUM COMPOUNDS, N.O.S. (BARIUM CHLORIDE), 6.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number10326-27-9
  • UN NumberUN1564
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Boiling Point C (° C)1560
  • Boiling Point F (° F)2840
  • Melting Point C (° C)963
  • Melting Point F (° F)1765.4
  • DescriptionBarium Chloride Dihydrate, ACS Reagent, Wide-Mouth Container, 100 g
Documents & Certificates
Reviews
Q&A