Reagents Boric Acid, Purified, Fiber Drum, 50 kg

Representative image only.

Reagents Boric Acid, Purified, Fiber Drum, 50 kg

Reagents | Mfr: C2104300-50F4| Item: UX-62283-41

Manufactured using the highest quality raw materials in ISO 9001 facility

$1,495.00USD / Each

$1,495.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.44
  • Melting point: 168 to 170 °C (334.4 to 338 °F)
  • Solubility information: 47.2 g/L at 20 °C (68 °F)
  • 50 kg fiber drum

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaH3BO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)50000
  • Formula Weight61.83
  • PackagingFiber drum
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10043-35-3
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)300
  • Boiling Point F (° F)572
  • Refractive Index1.44
  • DescriptionBoric Acid, Purified, Fiber Drum, 50 kg
Documents & Certificates
Reviews
Q&A