Reagents Boric Acid, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Boric Acid, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2104300-500A5| Item: UX-78910-48

Manufactured using the highest quality raw materials in ISO 9001 facility

$53.10USD / Each

$53.10USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.44
  • Melting point: 168 to 170 °C (334.4 to 338 °F)
  • Solubility information: 47.2 g/L at 20 °C (68 °F)
  • 500 g wide-mouth container
  • 20 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

20 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaH3BO3
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight61.83
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10043-35-3
  • Shelf Life (days)7325
  • Shelf Life (months)240
  • Boiling Point C (° C)300
  • Boiling Point F (° F)572
  • Refractive Index1.44
  • DescriptionBoric Acid, Purified, Wide-Mouth Container, 500 g
  • Warranty20 years
Documents & Certificates
Reviews
Q&A