Reagents Bromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Amber Glass Bottle, 100 mL

Representative image only.

Reagents Bromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Amber Glass Bottle, 100 mL

Reagents | Mfr: C7106300-100C| Item: UX-78910-56

Manufactured using the highest quality raw materials in ISO 9001 facility

$29.60USD / Each

$29.60USD / Each
Product Description

Key Features

  • Aqueous
  • Dark reddish-purple liquid
  • Density: 1
  • Boiling point: approximately 100 °C (212 °F)
  • Melting point: approximately 0 °C (32 °F)
  • Solubility information: miscible
  • pH range: 3.0 (yellow) to 4.6 (blue)
  • 100 mL amber glass bottle
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Two year warranty
Specifications
  • Concentration0.04% (w/v)
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)100
  • PackagingAmber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number62625-28-9, 7732-18-5
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionBromophenol Blue Indicator, 0.04 % (w/v), Volumetric, Amber Glass Bottle, 100 mL
  • Warranty2 year
Documents & Certificates
Reviews
Q&A