Reagents Calcium Chloride Dihydrate , 2.5kg, ACS Reagent

Representative image only.

Reagents Calcium Chloride Dihydrate , 2.5kg, ACS Reagent

Reagents | Mfr: C1163100-2.5A5| Item: UX-78919-59

Manufactured using the highest quality raw materials in ISO 9001 facility

$713.00USD / Each

$713.00USD / Each
Product Description

Key Features

  • Minimum Assay 74%
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCaCl2·2H2O
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight147.01
  • PackagingPoly Jar
  • CAS Number10035-04-8
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionCalcium Chloride Dihydrate, 2.5kg, ACS Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A