Reagents Calcium Chloride Dihydrate, Technical, Wide-Mouth Container, 500 g

Representative image only.

Reagents Calcium Chloride Dihydrate, Technical, Wide-Mouth Container, 500 g

Reagents | Mfr: C2163700-500A5| Item: UX-78910-71

Manufactured using the highest quality raw materials in ISO 9001 facility

$35.60USD / Each

$35.60USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.84
  • Boiling point: >1600 °C (>2912 °F)
  • Solubility information: 977 g/L at 0 °C (32 °F)
  • pH range: 4.5 to 8
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradeTechnical
  • Linear FormulaCaCl2·2H2O
  • ClassSalts
  • FormSolid
  • SynonymsCalcium chloride 2-hydrate
  • Weight (g)500
  • Formula Weight147.01
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number10035-04-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)176
  • Melting Point F (° F)348.8
  • DescriptionCalcium Chloride Dihydrate, Technical, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A