Reagents Chloroform, ACS Reagent, Amber Glass Bottle, 1 L

Representative image only.

Reagents Chloroform, ACS Reagent, Amber Glass Bottle, 1 L

Reagents | Mfr: C1175500-1C1| Item: UX-78918-59

Manufactured using the highest quality raw materials in ISO 9001 facility

$134.75USD / Each

$134.75USD / Each
Product Description

Key Features

  • Minimum Assay 99.8%
  • Colorless liquid
  • Density: 1.48
  • Boiling point: 60.5 to 61.2 °C (140.9 to 142.16 °F)
  • Solubility information: 8 g/L at 20 °C (68 °F)
  • 1 L amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaCHCl3
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsTCM, Trichloromethane, Methyl trichloride, Methane trichloride
  • Volume (mL)1000
  • Formula Weight119.38
  • Vapor Pressure (torr)158.263
  • PackagingAmber Glass Bottle, Safety Coated
  • DOT InformationCHLOROFORM, 6.1
  • CAS Number67-66-3
  • UN NumberUN1888
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-63.5
  • Melting Point F (° F)-82.3
  • Refractive Index1.446
  • DescriptionChloroform, 1L, ACS Reagent
  • Warranty5 years
Documents & Certificates
Reviews
Q&A