Reagents Conductivity Standard 10 µS/cm, 4L

Representative image only.

Reagents Conductivity Standard 10 µS/cm, 4L

Reagents | Mfr: CC900000-4A| Item: UX-78920-88

Manufactured using the highest quality raw materials in ISO 9001 facility

$185.00USD / Each

$185.00USD / Each

Buy It Together

Current Item

Reagents Conductivity Standard 10 µS/cm, 4L
Reagents Conductivity Standard 10 µS/cm, 4L
7892088

$185.00USD / Each
+ =
Total Price 205.10 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 10µS/cm, (9 to 11µS/cm)
  • 6 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

6 Month Warranty
Specifications
  • ClassConductivity
  • ChemistryStandards
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly Bottle
  • Shelf Life (days)210
  • Shelf Life (months)6
  • DescriptionConductivity Standard 10 µS/cm, 4L,
  • Warranty6 month
Documents & Certificates
Reviews
Q&A