Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Natural Poly Container, 125 g

Representative image only.

Reagents Cupric Sulfate Pentahydrate, ACS Reagent, Natural Poly Container, 125 g

Reagents | Mfr: C1190500-125A6| Item: UX-62281-67

Manufactured using the highest quality raw materials in ISO 9001 facility

$63.70USD / Each

$63.70USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Blue solid
  • Density: 2.29
  • Boiling point: >150 °C (302 °F)
  • Solubility information: 220 g/L at 25 °C (77 °F)
  • 125 g natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaCuSO4·5H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsCupric sulfate 5-hydrate, Copper (II) sulfate pentahydrate
  • Weight (g)125
  • Formula Weight249.69
  • PackagingNatural poly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7758-99-8
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)110
  • Melting Point F (° F)230
  • DescriptionCupric Sulfate Pentahydrate, ACS Reagent, Natural Poly Container, 125 g
Documents & Certificates
Reviews
Q&A