Reagents Cupric Sulfate Pentahydrate, Purified, Wide-Mouth Container, 500g

Representative image only.
Reagents Mfr # C2190500-500A5 – Item # EW-78911-04
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Blue solid
  • Density: 2.29
  • Boiling point: >150 °C (>302 °F), decomposes with loss of water
  • Melting point: 110 °C (230 °F), loses water
  • Solubility information: 220 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

$40.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Purity GradePurified
  • Linear FormulaCuSO4·5H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsCupric sulfate 5-hydrate, Copper (II) sulfate pentahydrate
  • Weight (g)500
  • Formula Weight249.69
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7758-99-8
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionCupric Sulfate Pentahydrate, Purified, Wide-Mouth Container, 500g
  • Warranty10 year
10 Year Warranty

Key Features

  • Blue solid
  • Density: 2.29
  • Boiling point: >150 °C (>302 °F), decomposes with loss of water
  • Melting point: 110 °C (230 °F), loses water
  • Solubility information: 220 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat