Reagents Cupriethylenediamine, 1.0M, Standardized, Amber Glass Bottle, 4 L

Representative image only.

Reagents Cupriethylenediamine, 1.0M, Standardized, Amber Glass Bottle, 4 L

Reagents | Mfr: C2191800-4C| Item: UX-78924-67

Manufactured using the highest quality raw materials in ISO 9001 facility

$454.00USD / Each

$454.00USD / Each
Product Description

Key Features

  • Cellulose solvent
  • Dark blue liquid
  • Density: 1.1
  • Solubility information: miscible
  • 4 L amber glass bottle
  • 1 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

1 Year Warranty
Specifications
  • ClassMixture
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingAmber glass bottle
  • DOT InformationCUPRIETHYLENEDIAMINE SOLUTION, 8 (6.1), P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1761
  • Shelf Life (days)395
  • Shelf Life (months)12
  • DescriptionCupriethylenediamine, 1.0M, Standardized, Amber Glass Bottle, 4 L
  • Warranty1 year
Documents & Certificates
Reviews
Q&A