Reagents Cyclohexane, ACS Reagent, Amber Glass Bottle, 4 L

Representative image only.

Reagents Cyclohexane, ACS Reagent, Amber Glass Bottle, 4 L

Reagents | Mfr: C1193500-4C| Item: UX-78921-04

Manufactured using the highest quality raw materials in ISO 9001 facility

$272.75USD / Each

$272.75USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless liquid
  • Density: 0.78
  • Solubility information: 0.05 g/L at 20 °C (68 °F)
  • 4 L amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaC6H12
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • SynonymsHexahydrobenzene, Hexamethylene
  • Volume (mL)4000
  • Formula Weight84.16
  • Vapor Pressure (torr)77.256
  • PackagingAmber glass bottle
  • DOT InformationCYCLOHEXANE, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number110-82-7
  • UN NumberUN1145
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)82.98
  • Boiling Point F (° F)181.36
  • Melting Point C (° C)-103.5
  • Melting Point F (° F)-154.3
  • Refractive Index1.426
  • DescriptionCyclohexane, ACS Reagent, Amber Glass Bottle, 4 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A