Reagents Deionized Water; MDA, APHA, EPA, AOAC Grade, Cubitainer® Package, 20 L

Representative image only.

Reagents Deionized Water; MDA, APHA, EPA, AOAC Grade, Cubitainer® Package, 20 L

Reagents | Mfr: C2346400-20F| Item: UX-78927-71

Manufactured using the highest quality raw materials in ISO 9001 facility

$130.00USD / Each

$130.00USD / Each
Product Description

Key Features

  • Water meets MDA, APHA, EPA, AOAC grade standards
  • Colorless liquid
  • Density: 1
  • Solubility information: miscible
  • 20 L Cubitainer package
  • 18 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

18 Month Warranty
Specifications
  • Purity GradeMDA, APHA, EPA, AOAC
  • Empirical FormulaH2O
  • ChemistrySolvents
  • FormLiquid
  • SynonymsDihydrogen oxide, Dihydrogen monoxide
  • Volume (mL)20000
  • Viscosity (cSt)0.8917
  • Formula Weight18.02
  • Vapor Pressure (torr)23.8
  • PackagingCubitainer
  • DOT InformationNot regulated by DOT
  • Storage SymbolAmbient
  • CAS Number7732-18-5
  • Shelf Life (days)540
  • Shelf Life (months)18
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionDeionized Water; MDA, APHA, EPA, AOAC Grade, Cubitainer® Package, 20 L
  • Warranty18 months
Documents & Certificates
Reviews
Q&A