Reagents Ethyl Alcohol, Denatured, 95%, Round Metal Can, 20 L

Representative image only.

Reagents Ethyl Alcohol, Denatured, 95%, Round Metal Can, 20 L

Reagents | Mfr: C2120500-20E6| Item: UX-78926-90

Manufactured using the highest quality raw materials in ISO 9001 facility

$497.00USD / Each

$497.00USD / Each
Product Description

Key Features

  • 190 proof
  • Colorless liquid
  • Density: 0.8
  • Boiling point: approximately 77 °C (170.6 °F)
  • Melting point: approximately -90 °C (-130 °F)
  • Flash point: 14 °C (57.2 °F)
  • Solubility information: miscible
  • 20 L round metal can
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Concentration0.95
  • ClassSolvents
  • Proof190
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)20000
  • Vapor Pressure (torr)48.4
  • PackagingRound metal can
  • DOT InformationALCOHOLS, N.O.S. (ETHANOL, METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number64-17-5
  • UN NumberUN1987
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionEthyl Alcohol, Denatured, 95%, Round Metal Can, 20 L
  • Warranty10 year
Documents & Certificates
Reviews
Q&A