Reagents Ethyl Anhydrous, Denatured, 20L

Representative image only.

Reagents Ethyl Anhydrous, Denatured, 20L

Reagents | Mfr: C2120600-20E6| Item: UX-78926-78

Manufactured using the highest quality raw materials in ISO 9001 facility

$549.00USD / Each

$549.00USD / Each
Product Description

Key Features

  • 200 Proof
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Linear FormulaCH3CH2OH
  • ClassSolvents
  • ChemistrySolvents
  • Proof200
  • FormLiquid
  • Volume (mL)20000
  • Formula Weight46.07
  • PackagingRieke Can
  • DOT InformationALCOHOLS, N.O.S. (ETHANOL, METHANOL, ISOPROPANOL), 3
  • CAS Number64-17-5
  • UN NumberUN1987
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionEthyl Anhydrous, Denatured, 20L,
  • Warranty10 year
Documents & Certificates
Reviews
Q&A