Reagents Ferrous Sulfate, ACS Reagent, Wide-Mounth Container, 500 g

Representative image only.

Reagents Ferrous Sulfate, ACS Reagent, Wide-Mounth Container, 500 g

Reagents | Mfr: C1207000-500A5| Item: UX-78911-40

Manufactured using the highest quality raw materials in ISO 9001 facility

$50.20USD / Each

$50.20USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Green or bluish-green solid
  • Density: 1.9
  • Boiling point: >300 °C (572 °F)
  • Solubility information: 295 g/L at 25 °C (77 °F)
  • 500 g wide-mounth container
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaFeSO4·7H2O
  • ClassSalts
  • SynonymsFerrous sulfate heptahydrate, Iron sulfate, Iron(II) sulfate heptahydrate
  • FormSolid
  • Weight (g)500
  • Formula Weight278.01
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7782-63-0
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)64
  • Melting Point F (° F)147.2
  • DescriptionFerrous Sulfate, ACS Reagent, Wide-Mounth Container, 500 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A