Reagents Formic Acid, 88%, ACS Reagent, Clear Glass Bottle, 2.5 L

Representative image only.

Reagents Formic Acid, 88%, ACS Reagent, Clear Glass Bottle, 2.5 L

Reagents | Mfr: C1105000-2.5D1| Item: UX-62281-19

Manufactured using the highest quality raw materials in ISO 9001 facility

$191.50USD / Each

$191.50USD / Each
Product Description

Key Features

  • Minimum Assay 88%
  • Colorless liquid
  • Density: 1.24
  • Flash point: 50 °C (122 °F)
  • Refractive index: 1.370 to 1.372nD
  • 2.5 L clear glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaHCOOH
  • ClassAcid
  • FormLiquid
  • Volume (mL)2500
  • Formula Weight46.03
  • PackagingClear glass bottle
  • DOT InformationFORMIC ACID, 8 (3), P.G. II 
  • Storage SymbolAmbient
  • UN NumberUN1779
  • CAS Number64-18-6
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)101
  • Boiling Point F (° F)213.8
  • DescriptionFormic Acid, 88%, ACS Reagent, Clear Glass Bottle, 2.5 L
Documents & Certificates
Reviews
Q&A