Reagents Hydrochloric Acid, 5.0N, 1L, Standardized

Representative image only.

Reagents Hydrochloric Acid, 5.0N, 1L, Standardized

Reagents | Mfr: CH101400-1A| Item: UX-78915-86

Manufactured using the highest quality raw materials in ISO 9001 facility

$58.40USD / Each

$58.40USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 2 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Two year warranty
Specifications
  • Normality5
  • ClassStandardized
  • FormLiquid
  • Volume (mL)1000
  • Vapor Pressure (torr)150
  • PackagingPoly Bottle
  • DOT InformationHYDROCHLORIC ACID SOLUTION, 8
  • UN NumberUN1789
  • Shelf Life (days)760
  • Shelf Life (months)24
  • DescriptionHydrochloric Acid, 5.0N, 1L, Standardized
  • Warranty2 year
Documents & Certificates
Reviews
Q&A