Reagents Hydrogen Peroxide, 35% (w/w), 1L, Technical

Representative image only.

Reagents Hydrogen Peroxide, 35% (w/w), 1L, Technical

Reagents | Mfr: CH302000-1A| Item: UX-78915-90

Manufactured using the highest quality raw materials in ISO 9001 facility

$79.40USD / Each

$79.40USD / Each
Product Description

Key Features

  • Assay 34 to 36% (w/w)
  • 18 month warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

18 Month Warranty
Specifications
  • ClassAqueous
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)1000
  • PackagingPoly Bottle
  • DOT InformationHYDROGEN PEROXIDE, AQUEOUS SOLUTIONS, 5.1 (8)
  • CAS Number7722-84-1
  • UN NumberUN2014
  • Shelf Life (days)570
  • Shelf Life (months)18
  • DescriptionHydrogen Peroxide, 35% (w/w), 1L, Technical
  • Warranty18 months
Documents & Certificates
Reviews
Q&A