Reagents Isopropanol, ACS Reagent, Cubitainer® Package, 20 L

Representative image only.

Reagents Isopropanol, ACS Reagent, Cubitainer® Package, 20 L

Reagents | Mfr: C1121500-20F| Item: UX-78928-12

Manufactured using the highest quality raw materials in ISO 9001 facility

$613.00USD / Each

$613.00USD / Each
Product Description

Key Features

  • Minimum Assay 99.5%
  • Colorless liquid
  • Density: 0.78
  • Flash point: 12 °C (53.6 °F)
  • Refractive index: 1.376 to 1.378nD
  • 20 L Cubitainer® package
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaCH3CHOHCH3
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Synonyms2-Propanol, Dimethyl carbinol, IPA, Isopropyl alcohol, Isopropanol, Rubbing alcohol
  • Volume (mL)20000
  • Formula Weight60.1
  • Vapor Pressure (torr)31.502
  • PackagingCubitainer
  • DOT InformationISOPROPANOL, 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1219
  • CAS Number67-63-0
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)82.5
  • Boiling Point F (° F)180.5
  • Melting Point C (° C)-90
  • Melting Point F (° F)-130
  • DescriptionIsopropanol, ACS Reagent, Cubitainer, 20 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A