Reagents Lead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Lead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C1229000-500C2| Item: UX-78918-79

Manufactured using the highest quality raw materials in ISO 9001 facility

$270.50USD / Each

$270.50USD / Each
Product Description

Key Features

  • Minimum 99%
  • Colorless/white solid
  • Density: 4.53
  • Melting point: 470 °C (878 °F), decomposes
  • Solubility information: 565 g/L at 20 °C (68 °F)
  • 500 g wide-mouth amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer® are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaPb(NO3)2
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight331.21
  • PackagingWide-mouth amber glass bottle
  • DOT InformationLEAD NITRATE, 5.1 (6.1), P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1469
  • CAS Number10099-74-8
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionLead Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty5 years
Documents & Certificates
Reviews
Q&A