Reagents Magnesium Chloride Hexahydrate, Technical, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Magnesium Chloride Hexahydrate, Technical, Natural Poly Container, 2.5 kg

Reagents | Mfr: C2238500-2.5A5| Item: UX-78919-20

Manufactured using the highest quality raw materials in ISO 9001 facility

$211.00USD / Each

$211.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.56
  • Solubility information: 560 g/L at 25 °C (77 °F)
  • 2.5 kg natural poly container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Purity GradeTechnical
  • Linear FormulaMgCl2·6H2O
  • ClassSalts
  • FormSolid
  • Weight (g)2500
  • Formula Weight203.3
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7791-18-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)117
  • Melting Point F (° F)242.6
  • DescriptionMagnesium Chloride Hexahydrate, Technical, Natural Poly Container, 2.5 kg
  • Warranty5 years
Documents & Certificates
Reviews
Q&A