Reagents Magnesium Chloride Hexahydrate, Technical, Wide-Mouth Container, 500 g

Representative image only.

Reagents Magnesium Chloride Hexahydrate, Technical, Wide-Mouth Container, 500 g

Reagents | Mfr: C2238500-500A5| Item: UX-62285-20

Manufactured using the highest quality raw materials in ISO 9001 facility

$80.10USD / Each

$80.10USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.56
  • Solubility information: 560 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeTechnical
  • Linear FormulaMgCl2·6H2O
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)500
  • Formula Weight203.3
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7791-18-6
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)117
  • Melting Point F (° F)242.6
  • DescriptionMagnesium Chloride Hexahydrate, Technical, Wide-Mouth Container, 500 g
Documents & Certificates
Reviews
Q&A