Reagents Mercuric Chloride, 100g, ACS Reagent

Representative image only.

Reagents Mercuric Chloride, 100g, ACS Reagent

Reagents | Mfr: C1245500-100C2| Item: UX-78911-98

Manufactured using the highest quality raw materials in ISO 9001 facility

$272.75USD / Each

$272.75USD / Each
Product Description

Key Features

  • Minimum Assay 99.5%
  • 7 year warranty

More About this Item

ACS Reagent Grade.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaHgCl2
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight271.5
  • Vapor Pressure (torr)1
  • PackagingAmber Jar
  • DOT InformationMERCURIC CHLORIDE, 6.1
  • CAS Number7487-94-7
  • UN NumberUN1624
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionMercuric Chloride, 100g, ACS Reagent
  • Warranty7 year
Documents & Certificates
Reviews
Q&A