Reagents Mercuric Sulfate, 100g, ACS Reagent

Representative image only.

Reagents Mercuric Sulfate, 100g, ACS Reagent

Reagents | Mfr: C1248000-100C2| Item: UX-78911-99

Manufactured using the highest quality raw materials in ISO 9001 facility

$211.00USD / Each

$211.00USD / Each
Product Description

Key Features

  • Powder, Minimum Assay 98%
  • 10 year warranty

More About this Item

ACS Reagent Grade.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaHgSO4
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight296.65
  • Vapor Pressure (torr)< 0.0075
  • PackagingAmber Jar
  • DOT InformationMERCURY SULFATE, 6.1
  • CAS Number7783-35-9
  • UN NumberUN1645
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionMercuric Sulfate, 100g, ACS Reagent
  • Warranty10 year
Documents & Certificates
Reviews
Q&A