Reagents Methyl Orange Indicator, 0.1% (w/v), Volumetric, Natural Poly Bottle, 500 mL

Representative image only.
Reagents Mfr # C6118000-500A – Item # UX-78912-10
Manufactured using the highest quality raw materials in ISO 9001 facility
  • Prepared in accordance with ASTM D974 procedures
  • Orange liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 3.2 (pink) to 4.4 (yellow)
  • 500 mL natural poly bottle
  • 3 year warranty

PRODUCT OPTIONS

$27.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Concentration0.1% (w/v)
  • Purity GradeASTM
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number547-58-0, 7732-18-5
  • Shelf Life (days)1125
  • Shelf Life (months)36
  • Boiling Point C (° C)100
  • Boiling Point F (° F)212
  • Melting Point C (° C)0
  • Melting Point F (° F)32
  • DescriptionMethyl Orange Indicator, 0.1% (w/v), Volumetric, Natural Poly Bottle, 500 mL
  • Warranty3 year
3 Year Warranty

Key Features

  • Prepared in accordance with ASTM D974 procedures
  • Orange liquid
  • Density: 1
  • Solubility information: miscible
  • pH range: 3.2 (pink) to 4.4 (yellow)
  • 500 mL natural poly bottle
  • 3 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat