Reagents Methyl Orange, Sodium Salt, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents Methyl Orange, Sodium Salt, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C6117500-25C1| Item: UX-62286-79

Manufactured using the highest quality raw materials in ISO 9001 facility

$38.80USD / Each

$38.80USD / Each
Product Description

Key Features

  • Orange-yellow powder
  • Density: 0.99
  • Melting point: >300 °C (572 °F), decomposes
  • Solubility information: 2 g/L at 25 °C (77 °F)
  • pH range: 3.2 (pink) to 4.4 (yellow)
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradeACS
  • Linear FormulaC14H14N3NaO3S
  • ClassIndicator
  • ChemistryAnalytical Reagents
  • FormSolid
  • Weight (g)25
  • PackagingWide-mouth amber glass bottle
  • DOT InformationDYES, SOLID, TOXIC, N.O.S. (METHYL ORANGE), 6.1, P.G. III
  • Storage SymbolAmbient
  • CAS Number547-58-0
  • UN NumberUN3143
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionMethyl Orange, Sodium Salt, ACS Reagent, Wide-Mouth Amber Glass Bottle, 25 g
Documents & Certificates
Reviews
Q&A