Reagents n-Butyl Acetate, Amber Glass Bottle, 4 L

Representative image only.

Reagents n-Butyl Acetate, Amber Glass Bottle, 4 L

Reagents | Mfr: C2156300-4C| Item: UX-78923-22

Manufactured using the highest quality raw materials in ISO 9001 facility

$256.00USD / Each

$256.00USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.88
  • Boiling point: 124 to 126 °C (255.2 to 258.8 °F)
  • Flash point: 22 °C (71.6 °F)
  • Refractive index: 1.393 to 1.395nD
  • Solubility information: 7 g/L at 20 °C (68 °F)
  • 4 L amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

5 Year Warranty
Specifications
  • Linear FormulaC6H12O2
  • ClassSolvents
  • ChemistrySolvents
  • FormLiquid
  • Volume (mL)4000
  • Formula Weight116.16
  • Vapor Pressure (torr)9.7508
  • PackagingAmber glass bottle
  • DOT InformationBUTYL ACETATES, 3, P.G. II
  • Storage SymbolAmbient
  • CAS Number123-86-4
  • UN NumberUN1123
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)-78
  • Melting Point F (° F)-108.4
  • Descriptionn-Butyl Acetate, Amber Glass Bottle, 4 L
  • Warranty5 years
Documents & Certificates
Reviews
Q&A