Reagents Nickel Sulfate Hexahydrate, 100g, Purified

Representative image only.

Reagents Nickel Sulfate Hexahydrate, 100g, Purified

Reagents | Mfr: C2256500-100C2| Item: UX-78912-31

Manufactured using the highest quality raw materials in ISO 9001 facility

$76.90USD / Each

$76.90USD / Each
Product Description

Key Features

  • Reagents and solutions to meet your lab and research needs
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNiSO4·6H2O
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight262.85
  • PackagingAmber Jar
  • CAS Number10101-97-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionNickel Sulfate Hexahydrate, 100g, Purified
  • Warranty10 year
Documents & Certificates
Reviews
Q&A