Reagents Oxalic Acid Dihydrate, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Oxalic Acid Dihydrate, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2110500-500A5| Item: UX-78912-45

Manufactured using the highest quality raw materials in ISO 9001 facility

$53.10USD / Each

$53.10USD / Each
Product Description

Key Features

  • Colorless/white crystals
  • Density: 1.65
  • Melting point: 104 to 106 °C (219.2 to 222.8 °F)
  • Solubility information: 140 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaC2H6O6·2H2O
  • ClassSalts
  • FormSolid
  • SynonymsOxalic acid 2-hydrate
  • Weight (g)500
  • Formula Weight162.1
  • Packagingwide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number6153-56-6
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionOxalic Acid Dihydrate, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A