Reagents Phenol (Liquefied), Purified, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Phenol (Liquefied), Purified, Amber Glass Bottle, 500 mL

Reagents | Mfr: C2269500-500C| Item: UX-78912-54

Manufactured using the highest quality raw materials in ISO 9001 facility

$223.00USD / Each

$223.00USD / Each
Product Description

Key Features

  • Colorless to slightly pink liquid
  • Density: 1.07
  • Melting point: 40 to 42 °C (104 to 107.6 °F)
  • Flash point: 79 °C (174.2 °F)
  • Solubility information: miscible
  • pH range: 3.5 to 4.2
  • 500 mL amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaC6H5OH
  • ClassPhenol
  • ChemistryAnalytical Reagents
  • FormLiquid
  • Volume (mL)500
  • Formula Weight94.11
  • Vapor Pressure (torr)16.55
  • PackagingAmber glass bottle
  • DOT InformationPHENOL SOLUTIONS, 6.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number108-95-2
  • UN NumberUN2821
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)182
  • Boiling Point F (° F)359.6
  • DescriptionPhenol (Liquefied), Purified, Amber Glass Bottle, 500 mL
  • Warranty5 years
Documents & Certificates
Reviews
Q&A