Reagents Phenolphthalein, 1% (w/v), Volumetric, Natural Poly Bottle, 500 mL

Representative image only.

Reagents Phenolphthalein, 1% (w/v), Volumetric, Natural Poly Bottle, 500 mL

Reagents | Mfr: C7270220-500A| Item: UX-78912-66

Manufactured using the highest quality raw materials in ISO 9001 facility

$40.40USD / Each

$40.40USD / Each
Product Description

Key Features

  • Colorless liquid
  • Density: 0.8
  • Boiling point: approximately 77 °C (170.6 °F)
  • Melting point: approximately -90 °C (-130 °F)
  • Flash point: 14 °C (57.2 °F)
  • Solubility information: miscible
  • pH range: 8.0 (colorless) to 10.0 (pink)
  • 500 mL natural poly bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Concentration1% w/v in 95% Ethanol
  • ClassIndicator
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)500
  • PackagingPoly bottle
  • DOT InformationALCOHOLS, N.O.S. (ETHANOL, METHANOL), 3, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1987
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • DescriptionPhenolphthalein, 1% (w/v), Volumetric, Natural Poly Bottle, 500 mL
  • Warranty5 years
Documents & Certificates
Reviews
Q&A