Reagents Phosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 500 mL

Representative image only.

Reagents Phosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 500 mL

Reagents | Mfr: C5101950-500C| Item: UX-78916-30

Manufactured using the highest quality raw materials in ISO 9001 facility

$109.50USD / Each

$109.50USD / Each
Product Description

Key Features

  • Minimum Assay 85%
  • Colorless liquid
  • Density: 1.69
  • Solubility information: miscible
  • pH range: <1
  • 500 mL amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaH3PO4
  • ClassAcid
  • FormLiquid
  • Volume (mL)500
  • Formula Weight98
  • Vapor Pressure (torr)0.03
  • PackagingAmber glass bottle
  • DOT InformationPHOSPHORIC ACID SOLUTION, 8, P.G. III
  • Storage SymbolAmbient
  • CAS Number7664-38-2
  • UN NumberUN1805
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)21
  • Melting Point F (° F)69.8
  • DescriptionPhosphoric Acid, 85% w/w, ACS Reagent, Amber Glass Bottle, 500 mL
  • Warranty5 years
Documents & Certificates
Reviews
Q&A