Reagents Potassium Chloride, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Chloride, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2280500-500A5| Item: UX-78912-91

Manufactured using the highest quality raw materials in ISO 9001 facility

$53.00USD / Each

$53.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 1.98
  • Solubility information: 347 g/L at 20 °C (68 °F)
  • pH range: 5.4 to 8.6
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaKCl
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight74.55
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7447-40-7
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Boiling Point C (° C)1413
  • Boiling Point F (° F)2575.4
  • Melting Point C (° C)771
  • Melting Point F (° F)1419.8
  • DescriptionPotassium Chloride, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A