Reagents Potassium Chromate, ACS Reagent, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Chromate, ACS Reagent, Wide-Mouth Container, 500 g

Reagents | Mfr: C1281000-500A5| Item: UX-78912-93

Manufactured using the highest quality raw materials in ISO 9001 facility

$263.75USD / Each

$263.75USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Yellow crystal
  • Density: 2.73
  • Solubility information: 650 g/L at 25 °C (77 °F)
  • pH range: 8.6 to 9.8
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

10 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaK2CrO4
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight194.19
  • PackagingWide-mouth container
  • DOT InformationTOXIC, N.O.S. (POTASSIUM CHROMATE), 5.1 (6.1), P.G. III
  • Storage SymbolAmbient
  • CAS Number7789-00-6
  • UN NumberUN3288
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)971
  • Melting Point F (° F)1779.8
  • DescriptionPotassium Chromate, ACS Reagent, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A