Reagents Potassium Fluoride, Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Potassium Fluoride, Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C2284000-100C2| Item: UX-78912-99

Manufactured using the highest quality raw materials in ISO 9001 facility

$38.00USD / Each

$38.00USD / Each
Product Description

Key Features

  • Colorless/white solid
  • Density: 2.48
  • Boiling point: approximately 1505 °C (2741 °F)
  • Solubility information: 923 g/L at 20 °C (68 °F)
  • 100 g wide-mouth amber glass bottle
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaKF
  • ClassSalts
  • FormSolid
  • Weight (g)100
  • Formula Weight58.1
  • PackagingWide-mouth amber glass bottle
  • DOT InformationPOTASSIUM FLUORIDE, SOLID, 6.1, P.G. III
  • Storage SymbolAmbient
  • UN NumberUN1812
  • CAS Number7789-23-3
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • Melting Point C (° C)859
  • Melting Point F (° F)1578.2
  • DescriptionPotassium Fluoride, Anhydrous, Purified, Wide-Mouth Amber Glass Bottle, 100 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A