Reagents Potassium Iodide, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Iodide, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2286000-500A5| Item: UX-78913-17

Manufactured using the highest quality raw materials in ISO 9001 facility

$407.50USD / Each

$407.50USD / Each
Product Description

Key Features

  • White solid
  • Density: 3.12
  • Boiling point: approximately 100 °C (212 °F)
  • Solubility information: 1450 g/L at 20 °C (68 °F)
  • pH range: 6 to 9.2
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaKI
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight166
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7681-11-0
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • Melting Point C (° C)680
  • Melting Point F (° F)1256
  • DescriptionPotassium Iodide, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A