Reagents Potassium Permanganate, 0.10N, 4L, Standardized, Natural Poly Container
Representative image only.
Reagents Potassium Permanganate, 0.10N, 4L, Standardized, Natural Poly Container
Reagents | Mfr: C7288510-4A| Item: UX-78921-84
Manufactured using the highest quality raw materials in ISO 9001 facility
$115.00USD / Each
$115.00USD / Each
Product Description
Key Features
  • N/10, Assay 0.0995 to 0.1005 Normal, Aqueous
  • 18 month warranty
More About this Item
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.
18 Month Warranty
Specifications
  • Normality0.1
  • ClassStandardized
  • ChemistrySolutions
  • FormLiquid
  • Volume (mL)4000
  • PackagingPoly Bottle
  • Shelf Life (days)570
  • Shelf Life (months)18
  • DescriptionPotassium Permanganate, 0.10N, 4L, Standardized
  • Warranty18 months