Reagents Potassium Permanganate, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Potassium Permanganate, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2288500-500A5| Item: UX-78913-29

Manufactured using the highest quality raw materials in ISO 9001 facility

$100.25USD / Each

$100.25USD / Each
Product Description

Key Features

  • Purple-black solid
  • Density: 2.7
  • Melting point: >240 °C (>464 °F)
  • Solubility information: 76 g/L at 25 °C (77 °F)
  • 500 g wide-mouth container
  • 10 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

10 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaKMnO4
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight158.03
  • PackagingWide-mouth container
  • DOT InformationPOTASSIUM PERMANGANATE, 5.1, P.G. II
  • Storage SymbolAmbient
  • UN NumberUN1490
  • CAS Number7722-64-7
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionPotassium Permanganate, Purified, Wide-Mouth Container, 500 g
  • Warranty10 year
Documents & Certificates
Reviews
Q&A