Reagents Potassium Phosphate Dibasic, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Representative image only.

Reagents Potassium Phosphate Dibasic, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g

Reagents | Mfr: C1289500-100C2| Item: UX-78913-31

Manufactured using the highest quality raw materials in ISO 9001 facility

$50.60USD / Each

$50.60USD / Each
Product Description

Key Features

  • Minimum Assay 98%
  • Colorless/white powder
  • Density: 2.3
  • Solubility information: 1600 g/L at 20 °C (68 °F)
  • 100 g wide-mouth amber glass bottle
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradeACS
  • Linear FormulaK2HPO4
  • ClassSalts
  • SynonymsDipotassium hydrogen phosphate
  • FormSolid
  • Weight (g)100
  • Formula Weight174.18
  • PackagingWide-mouth amber glass bottle
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7758-11-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)340
  • Melting Point F (° F)644
  • DescriptionPotassium Phosphate Dibasic, ACS Reagent, Wide-Mouth Amber Glass Bottle, 100 g
  • Warranty5 years
Documents & Certificates
Reviews
Q&A