Reagents Potassium Phosphate Monobasic, ACS Reagent, Natural Poly Container, 3 kg

Representative image only.

Reagents Potassium Phosphate Monobasic, ACS Reagent, Natural Poly Container, 3 kg

Reagents | Mfr: C1289000-3A5| Item: UX-62282-66

Manufactured using the highest quality raw materials in ISO 9001 facility

$294.00USD / Each

$294.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless/white crystals
  • Density: 2.34
  • Boiling point: 400 °C (752 °F), loses water and becomes metaphosphate
  • Solubility information: 250 g/L at 25 °C (77 °F)
  • pH range: 4.1 to 4.5
  • 3 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaKH2PO4
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • SynonymsPotassium dihydrogen phosphate
  • Weight (g)3000
  • Formula Weight136.09
  • Vapor Pressure (torr)7.3
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number7778-77-0
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)252.6
  • Melting Point F (° F)486.68
  • DescriptionPotassium Phosphate Monobasic, ACS Reagent, Natural Poly Container, 3 kg
Documents & Certificates
Reviews
Q&A