Reagents Silver Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Representative image only.

Reagents Silver Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g

Reagents | Mfr: C1303000-500C2| Item: UX-78918-98

Manufactured using the highest quality raw materials in ISO 9001 facility

$3,515.00USD / Each

$3,515.00USD / Each
Product Description

Key Features

  • Minimum Assay 99%
  • Colorless or white crystal
  • Density: 4.35
  • Boiling point: approximately 440 °C (824 °F)
  • Melting point: 207.9 to 209.8 °C (406.22 to 409.64 °F)
  • Solubility information: 2340 g/L at 25 °C (77 °F)
  • 500 g wide-mouth amber glass bottle
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaAgNO3
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight169.87
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSILVER NITRATE, 5.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number7761-88-8
  • UN NumberUN1493
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionSilver Nitrate, ACS Reagent, Wide-Mouth Amber Glass Bottle, 500 g
  • Warranty7 year
Documents & Certificates
Reviews
Q&A