Reagents Silver Nitrate, Purified, Wide-Mouth Amber Glass Bottle, 25 g

Representative image only.

Reagents Silver Nitrate, Purified, Wide-Mouth Amber Glass Bottle, 25 g

Reagents | Mfr: C2303000-25C1| Item: UX-62285-57

Manufactured using the highest quality raw materials in ISO 9001 facility

$284.25USD / Each

$284.25USD / Each
Product Description

Key Features

  • Colorless or white solid
  • Density: 4.35
  • Boiling point: approximately 440 °C (824 °F)
  • Melting point: 207.9 to 209.8 °C (406.22 to 409.64 °F)
  • Solubility information: 2160 g/L at 20 °C (68 °F)
  • 25 g wide-mouth amber glass bottle

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Purity GradePurified
  • Linear FormulaAgNO3
  • ClassSalts
  • ChemistryInorganics
  • FormSolid
  • Weight (g)25
  • Formula Weight169.87
  • PackagingWide-mouth amber glass bottle
  • DOT InformationSILVER NITRATE, 5.1, P.G. II
  • Storage SymbolAmbient
  • CAS Number7761-88-8
  • UN NumberUN1493
  • Shelf Life (days)3680
  • Shelf Life (months)120
  • DescriptionSilver Nitrate, Purified, Wide-Mouth Amber Glass Bottle, 25 g
Documents & Certificates
Reviews
Q&A