Reagents Sodium Acetate Anhydrous, Purified, Wide-Mouth Container, 500 g

Representative image only.

Reagents Sodium Acetate Anhydrous, Purified, Wide-Mouth Container, 500 g

Reagents | Mfr: C2306400-500A5| Item: UX-78913-51

Manufactured using the highest quality raw materials in ISO 9001 facility

$65.60USD / Each

$65.60USD / Each
Product Description

Key Features

  • Colorless/white powder
  • Density: 1.53
  • Solubility information: 504 g/L at 25 °C (77 °F)
  • pH range: 7 to 9.2
  • 500 g wide-mouth container
  • 5 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

5 Year Warranty
Specifications
  • Purity GradePurified
  • Linear FormulaNaC2H3O2
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight82.03
  • Vapor Pressure (torr)< 0.7501
  • PackagingWide-mouth container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number127-09-3
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Boiling Point C (° C)123
  • Boiling Point F (° F)253.4
  • Melting Point C (° C)324
  • Melting Point F (° F)615.2
  • DescriptionSodium Acetate Anhydrous, Purified, Wide-Mouth Container, 500 g
  • Warranty5 years
Documents & Certificates
Reviews
Q&A