Reagents Sodium Acetate Trihydrate, 500g, Purified

Representative image only.

Reagents Sodium Acetate Trihydrate, 500g, Purified

Reagents | Mfr: C2306500-500A5| Item: UX-78913-55

Manufactured using the highest quality raw materials in ISO 9001 facility

$58.40USD / Each

$58.40USD / Each
Product Description

Key Features

  • Crystal
  • 7 year warranty

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradePurified
  • Linear FormulaNaC2H3O2·3H2O
  • ClassSalts
  • FormSolid
  • Weight (g)500
  • Formula Weight136.08
  • PackagingPoly Jar
  • CAS Number6131-90-4
  • Shelf Life (days)2585
  • Shelf Life (months)84
  • DescriptionSodium Acetate Trihydrate, 500g, Purified
  • Warranty7 year
Documents & Certificates
Reviews
Q&A