Reagents Sodium Borate, ACS Reagent, Natural Poly Container, 2.5 kg

Representative image only.

Reagents Sodium Borate, ACS Reagent, Natural Poly Container, 2.5 kg

Reagents | Mfr: C1310500-2.5A5| Item: UX-62282-84

Manufactured using the highest quality raw materials in ISO 9001 facility

$186.00USD / Each

$186.00USD / Each
Product Description

Key Features

  • Minimum Assay 99.5%
  • White granules
  • Density: 1.73
  • Boiling point: approximately 320 °C (608 °F)
  • Solubility information: 31.7 g/L at 25 °C (77 °F)
  • pH range: 9.15 to 9.2
  • 2.5 kg natural poly container

More About this Item

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form. Reagents utlilizes the highest quality raw materials to meet of exceed the specifications of the American Chemical Society.

Specifications
  • Purity GradeACS
  • Linear FormulaNa2B4O7·10H2O
  • ClassSalts
  • ChemistryInorganics
  • SynonymsSodium borate decahydrate, Borax, Sodium tetraborate decahydrate
  • FormSolid
  • Weight (g)2500
  • Formula Weight381.37
  • PackagingPoly container
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • CAS Number1303-96-4
  • Shelf Life (days)1855
  • Shelf Life (months)60
  • Melting Point C (° C)75
  • Melting Point F (° F)167
  • DescriptionSodium Borate, ACS Reagent, Natural Poly Container, 2.5 kg
Documents & Certificates
Reviews
Q&A